HCL 1.5 ERBIUM
Catalog #: CA11017-798
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$146.85 to $576.51
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$72.65 to $846.50
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
$309.76 to $2,237.83