HCL 1.5 BERYLLIUM
Catalog #: CA11017-776
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
64,40 $ to 734,93 $
VWR® ARISTAR® Multi-Element ICP and ICP-MS Certified Reference Standards, Enhanced Packaging
Supplier: VWR International
154,78 $
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
558,88 $ to 2 230,71 $