Dispenser Refill Packs for Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Catalog #: CA30617-198
Supplier: Cytiva
Specifications
- Pore size:0.45 µm
- Description:Dispenser Refill Pack, GN-6 Metricel® Membrane
- Material:Mixed Cellulose Ester
- Cat. no.:CA30617-198
- Supplier No.:68121
Specifications
About this item
These membranes have been designed specifically for use with the Sentino® automatic filter dispenser.
- Ideal for high volume laboratories
- Eliminate the time and waste of individually sealed packs of membrane
Delivery: Each box contains two additional discs for loading the dispenser.
Customers Who Bought This Also Bought