Order Entry
ContactUsLinkComponent
AMINO ACID F DILUTION SOLVENT
Amino Acid F Dilution Solvent, for Rapid Liquid Silica, 475 ml, Hach
Catalog #: CA11017-074
Supplier:  Hach
CAS Number:  
AMINO ACID F DILUTION SOLVENT
Amino Acid F Dilution Solvent, for Rapid Liquid Silica, 475 ml, Hach
Catalog #: CA11017-074
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Heteropoly blue
  • Description:
    Amino Acid F Dilution Solvent, for Rapid Liquid Silica
  • Size:
    475 mL
  • Method:
    Heteropoly Blue
  • Cat. no.:
    CA11017-074
  • Range:
    3 - 1000 μg/L SiO₂
  • Supplier No.:
    2353011
  • No. of Tests:
    250

Specifications

About this item

For determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method

Hach method 8282 Range: 3 - 1000 μg/L SiO2. Used with Amino Acid F Reagent Powder (2651155). Rapid Liquid™ methods offer an economical way to analyze multiple samples and to measure trace concentrations. Requires Pour-Thru Cell.

Customers Who Bought This Also Bought