ContactUsLinkComponent
M-COLIBLUE24 PLASTIC AMPULES
m-ColiBlue24® Media, Hach
Catalog #: CA11016-726
Supplier:  Hach
CAS Number:  
M-COLIBLUE24 PLASTIC AMPULES
m-ColiBlue24® Media, Hach
Catalog #: CA11016-726
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    CA11016-726
  • Supplier No.:
    2608450
Specifications
About this item
In 24 hours, Hach's m-ColiBlue24® Broth give you simultaneous results for total coliforms and E. coli.
Use m-ColiBlue24® Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.
  • Simultaneous results
  • Ready-to-use
  • Evaluate total coliform and E. coli
Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media
Use m-ColiBlue24® Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.
m-ColiBlue24 should only be used with pads manufactured by the following companies/brands: Pall Corporation (e.g., Sterile Petri Dishes with Absorbant Pads), Private Label/OEM products manufactured by Pall Gelman, Sartorius (cellulose and glass fiber pads), and Sartorius BioSart products with the integrated Petri dish. No other pad is compatible with the product, and the use of any pad other than those recommended may result in unreliable, inaccurate data.
With ready-to-use m-ColiBlue24® Broth, you can simultaneously detect total coliforms and E. coli—in just 24 hours, easily read and interpret results, save money by eliminating special equipment and UV lamps, minimize background growth of non-coliform bacteria, reduce false positives and false negatives, plus get superior sensitivity down to 1 CFU/100 mL, and conduct emergency testing and routine monitoring.
Customers Who Bought This Also Bought