Test Kits, COD, TNT plus®
Catalog #: CA11014-646
Supplier: Hach
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Reactor digestion
- Description:COD TNTplus™, ULR (digestion required)
- Size:25 Tubes/pack
- Cat. no.:CA11014-646
- Range:1 - 60 mg/L COD
- Supplier No.:TNT820
Specifications
About this item
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are color coded by parameter and range
- No reagent blank necessary
TNTplus™ works only with DR 6000™ UV-VIS, DR 3900™ Benchtop, and DR 1900™ Portable spectrophotometers.
Customers Who Bought This Also Bought