Order Entry
ContactUsLinkComponent
OZONE ACCUVAC COLOR TEST KIT
AccuVac® Ozone Test Kit, Color Disc, Hach
Catalog #: CA11014-408
Supplier:  Hach
CAS Number:  
OZONE ACCUVAC COLOR TEST KIT
AccuVac® Ozone Test Kit, Color Disc, Hach
Catalog #: CA11014-408
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Indigo trisulfonate
  • Description:
    AccuVac® Ozone Test Kit, Color Disc, AV, CD Platform
  • Method:
    Color Disc/Indigo Trisulfonate
  • Cat. no.:
    CA11014-408
  • Range:
    0.05 - 1.50 mg/L O₃
  • Supplier No.:
    2518050
  • No. of Tests:
    25

Specifications

About this item

Fast, accurate visual method for determining ozone concentrations

This 25-test kit is used to control disinfectant levels in water, wastewater, and industrial water. AccuVac® vials ensure no mess and no measuring. Indigo trisulfonate method masks interferences from chlorine and other oxidizing agents, and it makes ozone analysis easier. Simple, low-cost color-disc method is the most accurate visual test kit method. This rugged test kit contains all of the required reagents and apparatus to perform analysis in the field.

Ordering information: Kit includes color disc, comparator box and blank vial, sample beaker, 25 Ozone High Range AccuVac Ampules, AccuVac Snapper, instructions and carrying case.
Customers Who Bought This Also Bought