To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Volume:1.0 - 5.0 ml
- Description:Pipette, Variable Volume
- Supplier No.:BBP065
Specifications
About this item
Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation.
Outstanding accuracy with adjustments in 50 L increments (0.05 ml).