Order Entry
ContactUsLinkComponent
STABLCAL CALIBRATION KIT
Stablcal® Turbidity Standards Calibration Kit, 2100Q Portable Turbidimeter, Hach
Catalog #: CA11014-240
Supplier:  Hach
CAS Number:  
STABLCAL CALIBRATION KIT
Stablcal® Turbidity Standards Calibration Kit, 2100Q Portable Turbidimeter, Hach
Catalog #: CA11014-240
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Turbidity:
    100 NTU
    800 NTU
    10 NTU
    20 NTU
  • Description:
    Stablcal® Turbidity Standards Calibration Kit, Sealed Vials
  • Size:
    4 Ampule
  • Concentration:
    20, 100, 800, 10 NTU (for verification)
  • Cat. no.:
    CA11014-240
  • For Use With:
    2100Q Portable Turbidimeter
  • Supplier No.:
    2971205

Specifications

About this item

Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter

-Precise, Pre-mixed Formazin Solutions
-Based on Formazin Primary Standards
-Enhanced Stability Saves Time and Labor
-Confidence in results

  • Precise, Pre-mixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results

Stablcal Calibration Standards Kit, for 2100Q Portable Turbidimeter

Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit contains one each of 20, 100, and 800 NTU Stablcal vials for calibration and one 10 NTU Stablcal vial for verification.

Customers Who Bought This Also Bought