ContactUsLinkComponent
TOT DISSOLVED IRON TEST STRIPS
Total Dissolved Iron Test Strips, Hach
Catalog #: CA11014-182
Supplier:  Hach
CAS Number:  
TOT DISSOLVED IRON TEST STRIPS
Total Dissolved Iron Test Strips, Hach
Catalog #: CA11014-182
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    CA11014-182
  • Range:
    0 - 5 mg/L
  • Supplier No.:
    2745325
  • No. of Tests:
    25
Specifications
About this item
In domestic water supplies, iron can do great economic damage by staining laundry and porcelain fixtures and by producing an off taste in beverages
Hach's test strips provide the easiest method for determining low to mid-range iron concentrations. Technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.
Hach's test strips provide the easiest method for determining low to mid-range iron concentrations. Technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.
In water supplies used for commercial beverage production, low levels of iron are critical.
Customers Who Bought This Also Bought