ContactUsLinkComponent
CLEANER ACID CITRAJET 1GAL CS4
Citrajet® Low-Foaming Liquid Acid Cleaners
Catalog #: CA47732-996
Supplier:  Alconox
CLEANER ACID CITRAJET 1GAL CS4
Citrajet® Low-Foaming Liquid Acid Cleaners
Catalog #: CA47732-996
Supplier:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    4×3.8 L (1 gal.)
  • Packaging:
    Bottle
  • pH:
    2.5
  • Supplier No.:
    2001
Specifications
About this item
Citrajet is a low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing.
  • Dilute 2:100
  • Ideal for use as acid wash or rinse
  • Concentrated, phosphate free, biodegradable and readily disposable
  • Combines acid and detergent ingredients to remove metal oxides, salts and inorganic residues
Low foaming acid cleaner and rinse for high performance results from your cleaning machinery, such as ultrasonic cleaners.
It can be used to clean laboratory ware, sampling apparatus, dairy equipment optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.
And it can be used to remove scale, metal oxides, metal complexes, trace inorganics, milkstone, soil, grit, buffing compound, grime, grease, fats, oils, particulates, amphoteric peptides, deposits, chemical and solvents.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.
Cleaning method: Laboratory ware washers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electro cleaning, ultrasonic cleaners flow through clean-in-place and manual cleaning if proper skin and eye protection is used.
Certifications: USDA compliant. FDA certified.
Customers Who Bought This Also Bought