SP Bel-Art Protective Tapes, Bel-Art Products, a part of SP
Catalog #: 30617-004
Supplier: Bel-Art Products, a Part of SP
Specifications
- Length:32.9 m
- Width:1½"
- Color:Clear
- Description:Protective Tape Rolls
- Supplier No.:F13491-0150
Specifications
About this item
Tapes made of PP provides great versatility for heavy-duty lab use.
Tapes protect labels from the freezer, autoclave, acids, alkalis and organic solvents.
Customers Who Bought This Also Bought