To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Bovine Serum Albumin
- Clonality:Polyclonal
- Gene ID:280717
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Antigen symbol:BSA
- Conjugation:Unconjugated
- ELISA:Yes
- Size:1 ml
- Western blot:Yes
- Storage buffer:Whole antiserum containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Highly purified Bovine Serum Albumin in Freund's adjuvant
- Tested applications:ELISA, WB
- Purification:Unpurified
- Cat. no.:89353-520
- Supplier No.:GTX77156
Specifications
About this item
Rabbit polyclonal antibody to BSA
For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0