Anti-EGFR Rabbit Polyclonal Antibody
89304-704
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Epidermal growth factor receptor
- Antigen symbol:EGFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1956
- Antigen synonyms:PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- Storage buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around aa.1068~1072 (R-Y-S-S-D) derived from Human EGFR.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:89304-704
- Supplier No.:GTX50426
Rabbit Polyclonal antibody to EGFR
Suggested starting dilutions are as follows. WB: 1:500~1:1000, IHC-P: 1:50~1:100, ICC/IF: 1:100~1:200. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Frequently Bought Together