Dropette® Disposable Transfer Pipettes, Simport
Catalog #: CA11014-048
Supplier: Simport Scientific
Specifications
- Capacity:6.0 ml
- Sterility:Non sterile
- Description:-
- Cat. no.:CA11014-048
- Packaging:Boxes of 500
- Overall Length:22.5 cm
- Bulb Draw:2.3 ml
- Supplier No.:P200-60
Specifications
About this item
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
- Can be sealed and refrigerated
- Will not shatter or break
- Non toxic and inert and could be used in liquid nitrogen
- Uniform drop size having safe transfer of fluids
- Temperature resistant down to –196 °C
- Seven models provide graduations
Rubber bulbs are attached with glass pipettes and molded from see-through low density polyethylene. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Inert to biological fluids and most acids. Can be gas (EtO) sterilized.
Ordering information: Choose between 12 very popular models, available in several sizes, tip designs, and lengths.
Customers Who Bought This Also Bought