Order Entry
ContactUsLinkComponent
Cellulose Extraction Thimbles 603 Cellulose thimbles, 22 x 80 mm - thickness 1.5 mm, 25/pk
Whatman™ Standard Cellulose Extraction Thimbles
Catalog #: CA11013-546
Supplier:  Cytiva
Cellulose Extraction Thimbles 603 Cellulose thimbles, 22 x 80 mm - thickness 1.5 mm, 25/pk
Whatman™ Standard Cellulose Extraction Thimbles
Catalog #: CA11013-546
Supplier:  Cytiva

Specifications

  • Description:
    Grade 603 Standard cellulose extraction thimble
  • Thickness:
    1.5 mm
  • Max. operating temperature:
    130 °C
  • I.D.× Body Length:
    22×80 mm
  • Supplier No.:
    10350211

Specifications

About this item

Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.

Customers Who Bought This Also Bought