ContactUsLinkComponent
Nucleic Acid Stain 1Ml
LabSafe™ Nucleic Acid Stain (10,000X), G-Biosciences
Catalog #: CA89233-908
Supplier:  G-Biosciences
CAS Number:  
Nucleic Acid Stain 1Ml
LabSafe™ Nucleic Acid Stain (10,000X), G-Biosciences
Catalog #: CA89233-908
Supplier:  G-Biosciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LabSafe™ Nucleic Acid Stain (10,000X)
  • Size:
    1 ml
  • Cat. no.:
    CA89233-908
  • Supplier No.:
    786-409
Specifications
About this item
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels.
  • Non-carcinogenic by the Ames-test
  • Used commonly in gel electrophoresis for visualization of nucleic acids
  • Replace toxic Ethidium Bromide (a potent mutagen)
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels. The results are negative in both the mouse marrow chromophilous erythrocyte micronucleus and mouse primary spermatocyte chromosomal aberration tests.
Customers Who Bought This Also Bought