Versaflo™ S-Series Headcovers, 3M™
Catalog #: CA89132-108
Supplier: 3M
Specifications
- Description:White Headcover
- Size:Medium-Large
- Cat. no.:CA89132-108
- Supplier No.:S-133L-5
Specifications
About this item
Fully assembled headcovers with integrated comfort suspension provide head and face coverage
Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or in situations where there is less soiling of the outer fabric.
The wide faceseal has soft elastic for improved comfort and reduced airflow noise. The exhalation panel vents the headcover without the traditional vent hole pattern or exhalation valve. The visor has an excellent field of view with reduced curvature for decreased reflections and glare.
Certifications: OSHA APF 25.
Customers Who Bought This Also Bought