ContactUsLinkComponent
Petriseal Clr, 0.50Inch 108Ft
Petriseal and Containerseal Tapes, Electron Microscopy Sciences
Catalog #: 102097-032
Petriseal Clr, 0.50Inch 108Ft
Petriseal and Containerseal Tapes, Electron Microscopy Sciences
Catalog #: 102097-032
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Clear
  • Description:
    Petriseal
  • Size:
    0.50"x108'
  • Cat. no.:
    102097-032
  • Supplier No.:
    70500-C
Specifications
About this item
Ideal sealing tapes for petri dishes, microplates, lids, caps, and other lab items.
  • Enjoy all the benefits of Petriseal and Containerseal in a new wider format
  • Containerseal XL makes it easier to seal large containers and bottles and to cover wider gaps
Stretches, conforms, and seals the irregular perimeter of microplates or petri dishes. Waterproof and chemical resistant. Tapes and adhesive resist most solvents and caustic agents. Temperature resistant, seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100°C.
Customers Who Bought This Also Bought