To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:50 ml
- Description:Biodegradable Reservoir
- Cat. no.:102096-998
- Supplier No.:70497-48
Specifications
About this item
Disposable reagent reservoirs.
- Biodegradable - Using a special additive to PET, these reservoirs will safely degrade over time
- 100% Eco-Friendly - Each case of Bio-Pure Reservoirs are packed in bio-degradable bags and recycled cardboard cases
A special additive helps the plastic bio-degrade by allowing bacteria and other organisms to breakdown the complex molecular chain. This speeds up the process to reduce the resin to its natural components within five years as opposed to 1,000 years or more.