To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Point Size:Extra Fine (0.6 mm)
- Color:Red
- Description:Solvent Resistant Pen
- Cat. no.:102093-906
- Supplier No.:62053-R
Specifications
About this item
Solvent Resistant Pens feature an ultra-permanent ink which is resistant to xylene, acetone, and formalin.
Markings will not run on embedding cassettes or frosted/painted slides and labware, even afterrepeated washes and solution treatments. The markings can be removed with acetic acid, an ethanol/xylene mixture or paraffin. Testing advised before use.