True North™ Cool Containers, Electron Microscopy Sciences
Catalog #: 102093-896
Supplier: Electron Microscopy Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Cool Containers, 1L Pan, Emerald
- Cat. no.:102093-896
- Supplier No.:62013-10
Specifications
About this item
These rigid single mold polyurethane containers provides durability for chilling your temperature sensitive samples. Excellent insulation properties for ultra colduse with ice, LN2, ice-salt slurries, and more.
- Convenient lid nests beneath bucket or pan and easily removes with comfortable recessed knobs
- Snug fit minimizes evaporation loss
- Handy pour spout for pouring melted ice, water, and solutions
- Ergonomic handles safely transport bucket or pan from work area to sink
- Deep internal cavity has enough room to add a rack
- Tapered sides make stacking and storage compact
- Available in a bucket or pan and in multiple sizes
Customers Who Bought This Also Bought