Specifications
- Description:Cardboard Freezer Box Divider, 81 Hole
- Cat. no.:102093-726
- Supplier No.:61615-80
Specifications
About this item
For use with stainless steel freezer racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes.
- Box footprint is 133×133 mm
Select a box and a partition that ft the size and number of vials for your needs. Cardboard boxes can be labelled with Cryo-Tags for easy identification.