Order Entry
ContactUsLinkComponent
Tellurite Tube Solution1% 5Ml Pk10
BBL™ Tellurite Solution 1%, BD Biosciences
Catalog #: CA11012-396
Supplier:  BD
CAS Number:  
BBL™ Tellurite Solution 1%, BD Biosciences
Catalog #: CA11012-396
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BBL™ Tellurite Solution 1%
  • Size:
    5 mL
  • Supplier No.:
    215294

Specifications

About this item

Tellurite solution is a sterile 1% solution of potassium tellurite, a differential agent.

  • Sterile
  • Recommended for the selective isolation of Staphylococci and Corynebacteria
  • 5 mL per vial

Coagulase-positive staphylococci reduce tellurite and produce black colonies.

Customers Who Bought This Also Bought