VWR® Precision Tweezers
Catalog #: 89259-986
Supplier: VWR International
Specifications
- Length:110 mm
- Description:Precision Tweezers, Style No. 5
- Material:Stainless steel
- Cat. no.:89259-986
Specifications
About this item
VWR® Precision Tweezers are stainless steel with very sharp, fine points.
- Anti-magnetic properties
- Resistant to acids
- Tweezers feature fine points
Precision tweezers are available in a range of lengths and order sizes, from individual to bulk.
Customers Who Bought This Also Bought