VWR® Filter Forceps
Catalog #: 89259-954
Supplier: VWR International
Specifications
- Length:114 mm (41/2")
- Material:Stainless steel
- Cat. no.:89259-954
Specifications
About this item
VWR® Filter Forceps are stainless steel with black polypropylene non-slip finger grips.
- Black polypropylene fabrication
- Non-slip finger grips
The tips are flat for handling membranes.
Customers Who Bought This Also Bought