VWR® Aluminum Test Tube Racks
Catalog #: 89259-898
Supplier: VWR International
Specifications
- Description:Test Tube Rack for 24 x 26 mm O.D. Tubes
- Material:Aluminium
- Cat. no.:89259-898
Specifications
About this item
VWR® Aluminum Test Tube Racks are punched from hard sheet aluminum.
- Edges and holes have a smooth finish
- Practical, rust-proof design
S-shaped rack is practical and rust-proof. The low profile and wide, flat base prevent rack from tipping. Rack also slides into other racks for compact storage.
Customers Who Bought This Also Bought