ContactUsLinkComponent
Crucible Tongs
VWR® Stainless Steel Crucible Holders, Corrugated Tip
Catalog #: 89259-892
Supplier:  VWR International
Crucible Tongs
VWR® Stainless Steel Crucible Holders, Corrugated Tip
Catalog #: 89259-892
Supplier:  VWR International
Specifications
  • Length:
    508 mm
  • Material:
    Stainless steel
  • Cat. no.:
    89259-892
Specifications
About this item
VWR® Stainless Steel Crucible Holders have tight-fitting corrugated tips for handling dishes in muffle furnaces.
  • Stainless steel construction
  • Corrugated tips
  • Joint for holding large-sized crucibles
They are constructed from a stainless steel rod with a diameter of 9 mm. There is a joint near the middle for holding large-sized crucibles.
Customers Who Bought This Also Bought