ContactUsLinkComponent
Neoprene Crucible Holders Filter Adapter
VWR® Neoprene Crucible Holders
Catalog #: 89259-864
Supplier:  VWR International
Neoprene Crucible Holders Filter Adapter
VWR® Neoprene Crucible Holders
Catalog #: 89259-864
Supplier:  VWR International
Specifications
  • Description:
    Neoprene Crucible Holder, Size 4, 30 mm height, 28 mm bottom O.D., 46 mm top O.D.
  • Cat. no.:
    89259-864
Specifications
About this item
These neoprene crucible holders are tapered, collar-type holders.
  • Used alone or with adjacent sizes
They support funnels, crucibles, and other equipment in vacuum-type filtration assemblies. They feature a positive, but easy-to-release gas-tight seal.
Customers Who Bought This Also Bought