VWR® Test Tube Baskets, Stainless Steel
Catalog #: 89259-824
Supplier: VWR International
Specifications
- Material:Stainless steel
- Cat. no.:89259-824
- L×D×H:30.5 x 30.5 x 6.4 cm (12 x 12 x 21/2")
Specifications
About this item
VWR® Stainless Steel Baskets are lightweight and strong.
- Resistant to acids and alkalis
- Stainless steel construction
- Lightweight
- Strong and durable
Test tube baskets will withstand acid and alkali solutions.
Customers Who Bought This Also Bought