Racks for Combitips Advanced® Pipette Tips, Eppendorf
Catalog #: CA89232-992
Supplier: Eppendorf
Specifications
- Description:Combitips advanced® Rack, for 8 Combitips advanced® (0.1 – 10 mL)
- Cat. no.:CA89232-992
- Supplier No.:0030089758
Specifications
About this item
Combitips® advanced Rack for 8 Combitips® advanced (0.1 - 10 ml), Eppendorf Quality™.
Customers Who Bought This Also Bought