Specifications
- Environmentally Preferable:Sustainable Packaging,Waste Reducing
- Size:5 L ( 1.32 gal.)
- Cat. no.:CA42000-000
- Packaging:Poly Bottle
- Supplier No.:1003
Specifications
About this item
Contrad 70 liquid detergent concentrate is phosphate-free, chlorine-free, and biodegradable
Cleans glass, plastic, ceramic, and ferrous metals by soaking, manual scrubbing, or ultrasonic cleaning. Contrad 70 is free-rinsing and disposable straight to the drain. Made with recycled packaging.