Order Entry
ContactUsLinkComponent
Sampling Spatula, 14In Sterileware
SP Bel-Art Non sterile and Sterileware® Sampling Spatulas, Bel-Art Products, a part of SP
Catalog #: 89236-110
Sampling Spatula, 14In Sterileware
SP Bel-Art Non sterile and Sterileware® Sampling Spatulas, Bel-Art Products, a part of SP
Catalog #: 89236-110

Specifications

  • Capacity:
    23 cm³
  • Length:
    35.6 cm (14")
  • Description:
    Sterileware® 14" Spatula
  • Blade Length:
    254 mm (10")
  • Handle Length:
    10.2 cm (4")
  • Supplier No.:
    369300050

Specifications

About this item

These V-shaped spatulas are ideal for every type of powdered or granulated material.

  • Available sterile or non sterile
  • Molded from FDA-grade materials
  • Safe for food, drug, and cosmetic contact
  • V shape is ideal for powdered or granulated material

The sturdy point is perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

Long 14" spatula is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄").

These one-piece spatulas are molded from FDA-grade materials and are safe for food, drug, and cosmetic contact.

Sterileware® Spatulas (9 and 14") are made of high-impact polystyrene, individually packaged, sterilized by gamma irradiation, lot stamped, and shipped with a Certificate of Processing. Non sterile 9" spatulas are made of polypropylene.

Customers Who Bought This Also Bought