Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Yellow,Blue
- Glove size:9
- Length:30.5 cm (12")
- Latex/Latex-free:Latex
- Thickness:27 mil
- EN norm:3121
- Cuff style:Pinked
- Liner material:Flock
- Finish:Diamond Embossed
- Cat. no.:CA32890-596
- Supplier No.:0022411090
Specifications
About this item
Gloves provide extra chemical protection and durability for more demanding applications due to a unique dipping process that combines neoprene over natural rubber latex for increased protection against a wide range of chemicals.
- Thick and rugged for excellent physical protection
- Patterned fingers provide added grip on wet and dry surfaces
- Worker comfort increased through sweat-wicking technology
Gloves are 27 millimeters thick for extra ruggedness, durability, and protection against physical hazards.
Use in the aerospace, automotive and transportation, chemical, and general handling industries. Uses include chemical processing; chemical plants and transportation; cleaning and de-greasing with soaps and detergent formulations; handling acids, alkalies, salts and ketones; handling specialty chemicals; pesticide manufacturing; and wiping down parts and machinery.
The diamond pattern provides added grip on wet or dry surfaces. Cotton flock lining absorbs perspiration and increases worker comfort.