Reagent alcohol absolute 94.0-96.0%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Catalog #: CAMK701910
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-17-5
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Size:4 L
- Supplier No.:7019-10
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Specially Denatured Alcohol Formula 3A (200 proof)(v/v):94.0 - 96.0 %
- Ethanol (v/v):88.5 - 92.5 %
- Methanol (v/v):3.5 - 5.5 %
- Isopropyl Alcohol (2-propanol)(v/v):4.0 - 6.0 %
- Total Alcohol (v/v):≥98.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Specific Gravity at 25°C:0.782 - 0.793
- Water (H₂O):≤0.5 %
- SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200:
- proof ethanol. Product Information (not specifications): Appearance:
- (clear, colorless liquid):
- Macron Fine Chemicals™, Reagent alcohol absolute 94.0-96.0%, AR® ACS (denatured with 5% isopropanol and 5% methanol):
Specification Test Results
About this item
Synonyms: Reagent alcohol
:Reagent alcohol
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:94,0-96,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
EINECS:200-578-6
UN:1987
:3
:II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought