Trichloroacetic acid (TCA) ≥99.0% (by acidimetry on anhydrous substance), crystals, AR® ACS, Macron Fine Chemicals™
Catalog #: CAMK292808
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 76-03-9
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber
- Size:125 g
- Cat. no.:CAMK292808
- Supplier No.:2928-08
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (Cl₃CCOOH) (dried basis)(by acidimetry):≥99.0 %
- ACS - Clarity of Solution:Passes Test
- ACS - Insoluble Matter:≤0.01 %
- ACS - Residue after Ignition:≤0.03 %
- ACS - Chloride (Cl):≤0.002 %
- ACS - Nitrate (NO₃):≤0.002 %
- ACS - Phosphate (PO₄):≤5 ppm
- ACS - Sulfate (SO₄):≤0.02 %
- ACS - Heavy Metals (as Pb):≤0.002 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- Macron Fine Chemicals™, Trichloroacetic acid (TCA) ≥99.0% (by acidimetry on anhydrous substance), crystals, AR® ACS:
Specification Test Results
About this item
Synonyms: TCA
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
Flash Pt:113 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 196 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 54…56 °C
:Flash Pt:
:Flash Pt: 113 °C (closed cup)
:M.W. 163.39 g/mol
Merck Index:12,09756
:Trichloroacetic acid (TCA)
EINECS:200-927-2
UN:1839
:8
:II
:113 °C
:GHS05
:GHS07
:GHS09
:GHS08
Storage Temperature:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought