VaporTrap™ Filter, Justrite®
Catalog #: 89232-200
Supplier: Justrite
Specifications
- Description:VaporTrap™ Filter
- Height:46"
- Cat. no.:89232-200
- Supplier No.:29916
Specifications
About this item
Filters reduces harmful VOC vapors found inside safety cabinets.
- Cylindrical design allows 360 degree air flow for maximum surface contact
- High strength magnets offer flexible placement anywhere within cabinet
Improve your work environment by reducing employee's inhalation exposure to harmful VOC's. They lower emissions of VOC's into the atmosphere.
Stainless steel wire mesh cartridge containing activated carbon adsorbs volatile organic chemical vapors.
Ordering information: Two filters per package.
Customers Who Bought This Also Bought