Specifications
- Description:Moisture Analyzer Test System (set of 12)
- Cat. no.:CA11010-734
- Supplier No.:30005792
Specifications
About this item
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
- Simply pour SmartCal™ into the sample pan and start
- Test takes just 10 minutes
- Easy to monitor and interpret instrument's performance over time
- Store results directly in the instrument or manually enter into a free validated Excel® report sheet
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.