Axygen® Deep Well and Assay Plates, Corning
Catalog #: 89230-176
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Well Volume:1.2 ml
- Color:Clear
- Description:Deep well plates
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:89230-176
- Packaging:Bulk
- Well Bottom Shape:Round
- Supplier No.:P-DW-12-HC-S
Specifications
About this item
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
- 0.6 ml deep well plate
- ANSI/SLAS 1-2004: Microplates - footprint dimensions
- Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
Customers Who Bought This Also Bought