Order Entry
ContactUsLinkComponent
Drierite Indicating 4 Mesh 2Kg
DRIERITE® Mesh Granules
Catalog #: CA71007-100
Supplier:  VWR
DRIERITE® Mesh Granules
Catalog #: CA71007-100
Supplier:  VWR
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Color:
    Blue
  • Description:
    DRIERITE® Indicating 4 Mesh
  • Size:
    2 kg
  • Cat. no.:
    CA71007-100
  • Packaging:
    Poly Bottle
  • Supplier No.:
    37226-380

Specifications

About this item

DRIERITE° will absorb 6 to 14% its weight in water from liquids or gases

It can be regenerated many times, and the granules do not become wet on saturation, nor do they change appearance or volume.

Customers Who Bought This Also Bought