VWR® Signature™ Frocks made with DuPont™ Tyvek® IsoClean® Material
Catalog #: 89127-312
Supplier: VWR International
Specifications
- Clothing Size:M
- Garment application:Laboratory
- Color:White
- Cat. no.:89127-312
Specifications
About this item
Frocks provide a high level of protection in cleanroom environments.
- High level of protection in cleanroom environments
- Chemically and biologically inert
- Low linting
- Static-free until laundered
These limited-use frocks are available in two styles. Frocks with covered zipper front feature tunneled elastic wrists and serged seams. These frocks are available with a bound neck. Frocks with five-snap front feature a bound neck and bound seams. All garments are pocketless.
Garments are chemically and biologically inert, low-linting, and static-free until laundered.
Customers Who Bought This Also Bought