VWR® Powder-Free Nitrile Examination Gloves
Catalog #: 82026-430
Supplier: VWR International
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue
- Glove size:XL
- Length:22.9 cm (9")
- Powdered/Powder-free:Powder-free
- Description:Nitrile Examination Gloves
- Disposable / reusable:Disposable
- Finger thickness:4.7 mil
- Palm thickness:3.9 mil
- Cuff thickness:3.1 mil
- Cat. no.:82026-430
Specifications
About this item
Ambidextrous gloves are extra soft and comfortable for every day use. They protect hands from a broad range of chemicals. Beaded cuff allows easy donning.
- Ambidextrous
- Nonsterile
- Extra soft and comfortable
- Protects hands from broad range of chemicals
- Powder-free
Ideal for use in industrial and laboratory applications
Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Textured gloves feature rolled, beaded cuffs.
Customers Who Bought This Also Bought