Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Catalog #: 10806-000
Supplier: Puritan Medical Products
Specifications
- Description:With adhesive
- Cat. no.:10806-000
- Handle Length:152.4 mm (6")
- Head Width:4.7 mm (0.188")
- Head Length:15.9 mm (0.625")
- Handle Width:2.5 mm (0.1")
- Supplier No.:806-WC
Specifications
About this item
These Puritan® swabs are ideal for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens.
Swabs with adhesive incorporate a nonsterile cotton tip secured by adhesive to the wooden handle. Swabs are also available without adhesive, for use where contamination of critical surfaces is a concern. Swabs are autoclavable.
They are also useful for cleaning delicate laboratory instruments.
Ordering information: Packaged in bags of 100 swabs, 10 bags to a pack, 10 packs to a case.
Customers Who Bought This Also Bought