ContactUsLinkComponent
EGM -2 Endothelail Cell Growth Medium-2 Singlequots Supplements And Growth Factors
SingleQuots® EGM®-2 Endothelial Kit
Catalog #: CA71005-532
CAS Number:  
EGM -2 Endothelail Cell Growth Medium-2 Singlequots Supplements And Growth Factors
SingleQuots® EGM®-2 Endothelial Kit
Catalog #: CA71005-532
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EGM®-2 Endothelial kit
  • Cat. no.:
    CA71005-532
  • Supplier No.:
    CC-4176
Specifications
About this item
EGM® -2 SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the EGM® -2 Endothelial Cell Growth Medium BulletKit® (Catalog Number CC-3162) which consists of basal medium and supplements packaged together.
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics® Primary cells.
The following Endothelial Cell Growth Medium BulletKits® are available:
EGM®-2 BulletKit® (CC-3162) – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
EGM® BulletKit® (CC-3124) – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
EGM®-Plus BulletKit® (CC-5035) – Enhanced version of EGM® Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM® Medium.
EGM®-2 MV BulletKit® (CC-3202) – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
Lonza guarantees the performance of Clonetics®/Poietics® Cells only if appropriate Clonetics®/Poietics® Media and Reagents are used exclusively and the recommended storage and use protocols are followed.
Delivery: Contents:
1 × Bottle FBS 10.00 ml
1 × Natural Cap Vial with Hydrocortisone 0.20 ml
1 × Gray Cap Vial with hFGF-B 2.00 ml
1 × White Cap Vial with VEGF 0.50 ml
1 × Yellow Cap Vial with R3-IGF-1 0.50 ml
1 × Blue Cap Vial with Ascorbic Acid 0.50 ml
1 × Green Cap Vial with hEGF 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Orange Cap Vial with Heparin 0.50 ml
Customers Who Bought This Also Bought