Orders LinkContactUsLinkComponent
EBM® Endothelial Cell Growth Basal Medium, Phenol Red Free
EBM® Endothelial cell growth basal medium, phenol red free
Catalog # CA71005-524
 :  CC-3129
EBM® Endothelial Cell Growth Basal Medium, Phenol Red Free
Catalog # CA71005-524
CAS Number:  
EBM® Endothelial Cell Growth Basal Medium, Phenol Red Free
EBM® Endothelial Cell Growth Basal Medium, Phenol Red Free
EBM® Endothelial cell growth basal medium, phenol red free
CAS Number:  
Price
$248.85
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
EBM® Phenol Red Free is an endothelial basal medium sold separately. This can be used with 1 kit of CC-4133 (endothelial cell growth medium SingleQuots® Supplements and Growth Factors for use with certain HUVEC cells).
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics® Primary Cells.
The following Endothelial Cell Growth Medium BulletKits® are available:
EGM® BulletKit® (CC-3124) – Optimized for HUVECs cat. no. CC-2517 and CC-2519, contains 2% FBS and BBE. Does not contain exogenous VEGF.
EGM®-Plus BulletKit® (CC-5035) – Enhanced version of EGM® Medium. Optimized for HUVECs cat. no. CC-2935. Contains 2% FBS and BBE. Does not contain exogenous VEGF. Faster proliferation than HUVECs isolated in EGM® Medium.
EGM®-2 BulletKit® (CC-3162) – Optimized for certain HUVECs, Aortic and Pulmonary artery endothelial cells. Contains 2% FBS and VEGF for rapid proliferation. Does not contain BBE.
EGM®-2 MV BulletKit® (CC-3202) – Coronary and Iliac artery, human microvascular endothelial cells, contains 5% FBS and VEGF for rapid proliferation. Does not contain BBE.
Lonza guarantees the performance of Clonetics®/Poietics® Cells only if appropriate Clonetics®/Poietics® Media and Reagents are used exclusively and the recommended storage and use protocols are followed.
 : Content: 1 × Bottle EBM® Phenol Free Basal Medium, 500 ml.
Specifications
Specifications
  • Description:
    EBM® Endothelial cell growth basal medium, phenol red free
Specifications