Order Entry
ContactUsLinkComponent
Blood Collection Tube 4ml lithium hep pk50
VACUETTE® Heparin Tubes, Greiner Bio-One
Catalog #: 95057-215
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Blood Collection Tube 4ml lithium hep pk50
VACUETTE® Heparin Tubes, Greiner Bio-One
Catalog #: 95057-215
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Ø:
    13 mm
  • Length:
    75 mm
  • Volume:
    3.5 ml
  • Description:
    VACUETTE® tube
  • Cat. no.:
    95057-215
  • Tube Dimensions:
    13 x 75 mm
  • Ring Color:
    Yellow
  • Supplier No.:
    454008

Specifications

About this item

Tubes feature an inner wall coated with spray-dried sodium heparin or lithium heparin.

VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with green non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.

  • Tubes contain anticoagulant
  • Color-coded rings in caps
  • PET plastic construction is virtually unbreakable

Tubes feature an inner wall coated with spray-dried sodium heparin or lithium heparin.

The anticoagulant heparin activates antithrombins, which block the coagulation cascade and produce a whole blood/plasma sample rather than clotted blood plus serum

VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with green non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.

Tubes are also available with lithium heparin coating and a gel separator. The specific gravity of the gel lies between the blood cells and plasma so that, during centrifugation, the gel barrier moves to form a stable barrier separating plasma from cells. Plasma may then be aspirated directly from the collection tube, eliminating the need for transfer to another container.

Customers Who Bought This Also Bought