Sodium dodecyl sulfate (SDS) ≥85.0% (by titrimetric analysis)
Catalog #: TCD0996-025G
Supplier: TCI America
CAS Number: 151-21-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:25 g
- Cat. no.:TCD0996-025G
- Supplier No.:D0996-25G
Specifications
Specification Test Results
- Purity/Analysis Method:85.0% (T)
Specification Test Results
About this item
Synonyms: Sodium lauryl sulfate,Dodecyl sulfate sodium salt,Lauryl sulfate sodium salt,Dodecyl sulfuric acid sodium salt,Dodecyl sulphate sodium salt,Sodium dodecyl sulfate,Dodecyl sulphuric acid sodium salt,Lauryl sulphate sodium salt
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
CAS Number:151-21-3
:Sodium dodecyl sulfate (SDS)
:Storage Temperature: Ambient
Melting Pt:207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
ADR:4.1,II
:Length
:Width
:Height
:Diameter
:Thickness
:85,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 207 °C
:Flash Pt:
:Flash Pt: >100 °C
:M.W. 288.38 g/mol
Merck Index:13,08710
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
:4.1
:II
:≥ 100 °C
:GHS02
:GHS06
:12,08782
:14,08636
Customers Who Bought This Also Bought