Ammonia solution 28.0 - 30.0% (NH₃ basis), BAKER ANALYZED® ACS, J.T.Baker®
Catalog #: JT9721-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1336-21-6
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear in a single shipper
- Size:500 ml
- Supplier No.:9721-02
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For laboratory, research or manufacturing use.:
- Appearance (colorless and free from suspended matter or sediment):Passes Test
- Assay (as NH₃):28.0 - 30.0%
- Color (APHA):≤5
- Specific Gravity at 60/60 °F:0.896 - 0.902
- Residue after Ignition:≤0.0020%
- Carbon Dioxide (CO₂):≤0.002%
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.5 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤2 ppm
- Trace Impurities - Aluminum (Al):≤200.0 ppb
- Arsenic and Antimony (as As):≤3000.0 ppb
- Trace Impurities - Barium (Ba):≤300.0 ppb
- Trace Impurities - Boron (B):≤50 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Gold (Au):≤200.0 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤200.0 ppb
- Trace Impurities - Magnesium (Mg):≤200.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Zinc (Zn):≤100.0 ppb
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Ammonia solution 28.0 - 30.0% (NH₃ basis), BAKER ANALYZED® ACS:
Specification Test Results
About this item
Synonyms: Ammonia aqueous,Ammonium hydroxide solution,Ammonium hydroxide
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 32…37.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –60…–57.5 °C
:Flash Pt:
:Flash Pt:
:M.W. 35.05 g/mol
:Ammonia solution
EINECS:215-647-6
:32…37.7 °C (1013 hPa)
Storage Temperature:Ambient
UN:2672
:8
:III
:GHS05
:GHS07
:GHS09
:M.O.C. Category NI
Customers Who Bought This Also Bought