EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL), J.T.Baker®
Catalog #: JT5648-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 6381-92-6
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE bottle, narrow mouth
- Size:1 L
- Cat. no.:JT5648-2
- Supplier No.:5648-02
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Titer, mg/mL, For Information Only:
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Each lot of this product is standardized at 25°C against NIST StandardReference Material Calcium Carbonate (Hydroxy Naphthol Blue endpoint).:
- SRM No…:
- The titer found falls within the range 0.980-1.020 mg CaCO₃/mL.:
- J.T.Baker®, EDTA disodium salt solution (1 ml = 1 mg CaCO₃), BAKER ANALYZED® for stable isotope labelling (SIL):
Specification Test Results
About this item
Synonyms: Ethylenedinitrilotetra-acetic acid disodium salt dihydrate,Kestranal® 2S 2 H2O,Edetic acid disodium salt dihydrate,Titriplex III,Disodium dihydrogen ethylenediaminetetraacetate dihydrate,Ethylenediaminetetra-acetic acid disodium salt dihydrate,Kestranal® 2S dihydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 372.24 g/mol
:M.O.C. Category NI
Customers Who Bought This Also Bought