Polysorbate 20 NF, Multi-Compendial, J.T.Baker®
Catalog #: JT4116-4
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 9005-64-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:200 ml
- Cat. no.:JT4116-4
- Supplier No.:4116-04
Specifications
Specification Test Results
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol :≤ 0.10 %
- Vegetable Based:
- This product utilizes ingredients of non-animal origin and non-peanut origin.:
- Suitable for use in injectable dosage forms.:
- Only Class 2 (1,4 Dioxane, Ethylene Glycol) and Class 3 (2-propanol) solvents are likely to be present. Class 2 solvents are below the Option 1 limits and any Class 3 solvent is <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:
- GMP Manufactured Product:
- Meets JPE Specifications:
- Meets N.F. Requirements:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- Store in airtight container, protected from light:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Acid Value:≤ 2.0
- NF - Residue on Ignition:≤ 0.25 %
- NF - Dioxane:≤ 10 ppm
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Hydroxyl Value, mg KOH/g:96 - 108
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Saponification Value, mg KOH/g:40 - 50
- NF - Caproic Acid (Composition of Fatty Acids):≤ 1.0 %
- NF - Caprylic Acid (Composition of Fatty Acids):≤ 10.0 %
- NF - Capric Acid (Composition of Fatty Acids):≤ 10.0 %
- NF - Lauric Acid (Composition of Fatty Acids):40.0 - 60.0 %
- NF - Myristic Acid (Composition of Fatty Acids):14.0 - 25.0 %
- NF - Palmitic Acid (Composition of Fatty Acids):7.0 - 15.0 %
- NF - Stearic Acid (Composition of Fatty Acids):≤ 11.0 %
- NF - Oleic Acid (Composition of Fatty Acids):≤ 11.0 %
- NF - Linoleic Acid (Composition of Fatty Acids):≤ 3.0 %
- NF - Peroxide Value:≤ 5.0
- NF - Water (H₂O):≤ 3.0 %
- Endotoxin Concentration (EU/mL):≤ 10
- EP/BP - Acid Value:≤ 2.0
- EP/BP - Total Ash:≤ 0.25 %
- EP/BP - Composition of Fatty Acids - Caproic Acid:≤ 1.0 %
- EP/BP - Composition of Fatty Acids - Caprylic Acid:≤ 10.0 %
- EP/BP - Composition of Fatty Acids - Capric Acid:≤ 10.0 %
- EP/BP - Composition of Fatty Acids - Lauric Acid:40.0 - 60.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:14.0 - 25.0 %
- EP/BP -Composition of Fatty Acids - Palmitic Acid:7.0 - 15.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 7.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≤ 11.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 3.0 %
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP/BP - Hydroxyl Value:96 - 108
- EP - Peroxide Value:≤ 10.0
- EP - Saponification Value:40 - 50
- EP - Water (H₂O):≤ 3.0 %
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- Appearance:Passes Test
- pH of 5% Solution at 25°C:5.0 - 7.0
- Arsenic (As):≤ 1.000 ppm
- Peroxide Value, meqO₂/kg:≤ 2.0
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Escherichia Coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- JPE - Identification 1:Passes Test
- JPE - Identification 2:Passes Test
- JPE - Identification 3:Passes Test
- JPE - Moisture Content:≤ 3.0 %
- JPE - Acid Value:≤ 4.0
- JPE - Saponification Value (mg KOH/g):43 - 55
- JPE - pH:4.0 - 7.0
- JPE - Residue on Ignition:≤ 0.25 %
- JPE - Specific Gravity at 20°C:1.090 - 1.130
- JPE - Viscosity (mm²/S):350 - 550
- JPE - Heavy Metals (as Pb):≤ 20 ppm
- JPE - Arsenic (As):≤ 2 ppm
- J.T.Baker®, Polysorbate 20 NF, Multi-Compendial:
Specification Test Results
About this item
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate,Sorbitan monolaurate
Density:1.11 g/cm³
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Ambient
EINECS:500-018-3
:M.O.C. Category R
Customers Who Bought This Also Bought